AMBH303C5318 Ambeed, Inc.

6-Amino-2-cyclopropylpyrimidin-4-ol

  • MDL number MFCD16988316
  • Cas Number : 954388-42-2
  • Molecular Weight : 151.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 122.2
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 278.5�0�t 760 mmHg
Product Link https://www.ambeed.com/products/954388-42-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/954388-42-2.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart