AMBH303C5375 Ambeed, Inc.

4-(Dimethylamino)-3-methylbenzaldehyde

  • MDL number MFCD01313806
  • Cas Number : 1424-69-7
  • Molecular Weight : 163.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1424-69-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1424-69-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart