AMBH303C5385 Ambeed, Inc.

1-Methyl-2-(methylsulfonyl)-5-nitro-1H-imidazole

  • MDL number MFCD26407320
  • Cas Number : 1615-53-8
  • Molecular Weight : 205.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1615-53-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1615-53-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart