AMBH303C53AB Ambeed, Inc.

(R)-1,1,1-Trifluoro-N-(pyrrolidin-2-ylmethyl)methanesulfonamide

  • MDL number MFCD11656044
  • Cas Number : 1186049-30-8
  • Molecular Weight : 232.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210
Purity 97
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1186049-30-8.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1186049-30-8.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart