AMBH303C5416 Ambeed, Inc.

Ethyltriphenylphosphonium acetate

  • MDL number MFCD00061389
  • Cas Number : 35835-94-0
  • Molecular Weight : 350.3906

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 6570
Storage temp. Inert atmosphere,Room Temperature
Physical Form liquid
Shipping Temperature Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/35835-94-0.html

Safety Information

signalword Danger
pcodes P210-P261-P280-P301+P310-P305+P351+P338-P312
MSDS https://www.ambeed.com/products/35835-94-0.html
pictograms GHS02,GHS06
hstatements H224-H300-H315-H319-H330-H335
Availability details for
Quantity
  • Availability
  • Add to Cart