AMBH303C5419 Ambeed, Inc.

SF1670

  • MDL number MFCD06411448
  • Cas Number : 345630-40-2
  • Molecular Weight : 307.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-(9,10-Dioxo-9,10-dihydrophenanthren-2-yl)pivalamide
Purity 98
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/345630-40-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/345630-40-2.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart