AMBH303C5441 Ambeed, Inc.

(S)-2-(2-Bromophenyl)pyrrolidine hydrochloride

  • MDL number MFCD12910614
  • Cas Number : 1224945-39-4
  • Molecular Weight : 262.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1224945-39-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1224945-39-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart