AMBH303C5452 Ambeed, Inc.

9-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-9H-carbazole

  • MDL number MFCD22415352
  • Cas Number : 870119-58-7
  • Molecular Weight : 369.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/870119-58-7.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/870119-58-7.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart