AMBH303C5460 Ambeed, Inc.

2-([1,1'-Biphenyl]-4-yl)-5,6-dihydroimidazo[2,1-b]thiazole hydrobromide

  • MDL number MFCD07281912
  • Cas Number : 1049780-58-6
  • Molecular Weight : 359.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1049780-58-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1049780-58-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart