AMBH303C5471 Ambeed, Inc.

3,3',3'',3'''-(Ethane-1,2-diylbis(azanetriyl))tetrakis(propan-1-ol)

  • MDL number MFCD23135560
  • Cas Number : 5261-23-4
  • Molecular Weight : 292.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/5261-23-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/5261-23-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart