AMBH303C548E Ambeed, Inc.

2-(2-Methoxyphenoxy)-1-(4-methoxyphenyl)propane-1,3-diol

  • MDL number MFCD29905047
  • Cas Number : 92409-15-9
  • Molecular Weight : 304.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 256.7
Storage temp. Sealed in dry,Store in freezer, under -20
Physical Form White to yellow powder or crystals or liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 500.8�t 760 mmHg
Product Link https://www.ambeed.com/products/92409-15-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/92409-15-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart