AMBH303C5490 Ambeed, Inc.

Ertapenem sodium

  • MDL number MFCD09475566
  • Cas Number : 153773-82-1
  • Molecular Weight : 497.50

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Sodium (4R,5S,6S)-3-(((3S,5S)-5-((3-carboxyphenyl)carbamoyl)pyrrolidin-3-yl)thio)-6-((R)-1-hydroxyethyl)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate
Purity 97
Flash Point 446°C
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20°C
Physical Form solid
Shipping Temperature Shipped in cold pack
Country of Origin China
Boiling point 813.9°C at 760 mmHg
Product Link https://www.ambeed.com/products/153773-82-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/153773-82-1.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart