AMBH303C549F Ambeed, Inc.

Sodium 2-((2,6-dichloro-3-methylphenyl)amino)benzoate

  • MDL number MFCD00077376
  • Cas Number : 6385-02-0
  • Molecular Weight : 318.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/6385-02-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/6385-02-0.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart