AMBH303C54A4 Ambeed, Inc.

3-Bromopropyl acetate

  • MDL number N/A
  • Cas Number : 592-33-6
  • Molecular Weight : 181.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 95
Storage temp. Inert atmosphere,2-8°C
Physical Form liquid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/592-33-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/592-33-6.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart