AMBH303C54B7 Ambeed, Inc.

Methyl 4-amino-6-ethoxypicolinate

  • MDL number MFCD16660808
  • Cas Number : 894803-87-3
  • Molecular Weight : 196.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/894803-87-3.html

Safety Information

signalword Warning
pcodes P264-P270-P301+P312-P330
MSDS https://www.ambeed.com/products/894803-87-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart