AMBH303C54E3 Ambeed, Inc.

Spiro[cyclopropane-1,3'-indolin]-2'-one

  • MDL number MFCD11934419
  • Cas Number : 13861-75-1
  • Molecular Weight : 159.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/13861-75-1.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/13861-75-1.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart