AMBH303C54F8 Ambeed, Inc.

2-(2-Bromoethyl)benzonitrile

  • MDL number MFCD24462563
  • Cas Number : 101860-81-5
  • Molecular Weight : 210.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 95
Storage temp. Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/101860-81-5.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/101860-81-5.html
pictograms GHS07
hstatements H302-H312-H332
Availability details for
Quantity
  • Availability
  • Add to Cart