AMBH303C54FF Ambeed, Inc.

2-(Carboxymethyl)-5-fluorobenzoic acid

  • MDL number MFCD19370105
  • Cas Number : 583880-95-9
  • Molecular Weight : 198.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/583880-95-9.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/583880-95-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart