AMBH303C5529 Ambeed, Inc.

Fenoprofen Calcium

  • MDL number MFCD00079289
  • Cas Number : 34597-40-5
  • Molecular Weight : 522.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Calcium 2-(3-phenoxyphenyl)propanoate
Purity 98
Flash Point 141.7°C
Storage temp. Inert atmosphere,Store in freezer, under -20°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 381.3°C at 760 mmHg
Product Link https://www.ambeed.com/products/34597-40-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/34597-40-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart