AMBH303C553F Ambeed, Inc.

Perfluorophenyl 5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoate

  • MDL number MFCD09952636
  • Cas Number : 120550-35-8
  • Molecular Weight : 410.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 289.6
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Shipped in cold pack
Country of Origin CN
Boiling point 555.3�0�t 760 mmHg
Product Link https://www.ambeed.com/products/120550-35-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/120550-35-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart