AMBH303C554D Ambeed, Inc.

(1S)-4,7,7-Trimethyl-3-oxo-2-oxabicyclo[2.2.1]heptane-1-carbonyl chloride

  • MDL number MFCD00135626
  • Cas Number : 39637-74-6
  • Molecular Weight : 216.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71-73
Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/39637-74-6.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
hcodes 8
MSDS https://www.ambeed.com/products/39637-74-6.html
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart