AMBH303C5567 Ambeed, Inc.

3-(3-Methoxyphenyl)-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD03030351
  • Cas Number : 890591-64-7
  • Molecular Weight : 218.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/890591-64-7.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/890591-64-7.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart