AMBH303C5570 Ambeed, Inc.

1-Boc-3-(5-Bromopyrimidin-2-yloxy)pyrrolidine

  • MDL number MFCD08460369
  • Cas Number : 914347-79-8
  • Molecular Weight : 344.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/914347-79-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/914347-79-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart