AMBH303C5579 Ambeed, Inc.

tert-Butyl 3-(2-(tert-butoxy)-2-oxoethyl)-4-oxopiperidine-1-carboxylate

  • MDL number MFCD11111810
  • Cas Number : 1010814-94-4
  • Molecular Weight : 313.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1010814-94-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1010814-94-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart