AMBH303C5596 Ambeed, Inc.

6-Nitro-4-(trifluoromethyl)quinolin-2(1H)-one

  • MDL number MFCD12828492
  • Cas Number : 328956-38-3
  • Molecular Weight : 258.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 284-285
Purity 97
Flash Point 176.8
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 368.7�t 760 mmHg
Product Link https://www.ambeed.com/products/328956-38-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/328956-38-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart