AMBH303C559B Ambeed, Inc.

2-Amino-2-(3-chloropyridin-4-yl)ethanol hydrochloride

  • MDL number MFCD30186145
  • Cas Number : 1956389-98-2
  • Molecular Weight : 209.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1956389-98-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1956389-98-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart