AMBH303C55AA Ambeed, Inc.

5-Fluoro-2-hydroxy-3-nitrobenzoic acid

  • MDL number MFCD18826200
  • Cas Number : 130046-91-2
  • Molecular Weight : 201.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/130046-91-2.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/130046-91-2.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart