AMBH303C55D5 Ambeed, Inc.

JNJ-40346527

  • MDL number MFCD30489233
  • Cas Number : 1142363-52-7
  • Molecular Weight : 461.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Cyano-N-(2-(4,4-dimethylcyclohex-1-en-1-yl)-6-(2,2,6,6-tetramethyltetrahydro-2H-pyran-4-yl)pyridin-3-yl)-1H-imidazole-2-carboxamide
Purity 97
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1142363-52-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1142363-52-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart