AMBH303C55E1 Ambeed, Inc.

N,N-Dipropyldinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine

  • MDL number MFCD27920535
  • Cas Number : 802902-36-9
  • Molecular Weight : 415.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/802902-36-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/802902-36-9.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart