AMBH303C55EA Ambeed, Inc.

Ethyl 2,3-dibromo-3-methylbutanoate

  • MDL number MFCD23135641
  • Cas Number : 79405-51-9
  • Molecular Weight : 287.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/79405-51-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/79405-51-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart