AMBH303C55EB Ambeed, Inc.

2-(3'-Chloro-[1,1'-biphenyl]-4-yl)-4,6-diphenyl-1,3,5-triazine

  • MDL number MFCD28975107
  • Cas Number : 1443049-84-0
  • Molecular Weight : 419.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1443049-84-0.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/1443049-84-0.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart