AMBH303C5648 Ambeed, Inc.

5-Methyl-7H-pyrrolo[2,3-d]pyrimidine

  • MDL number MFCD27987375
  • Cas Number : 1638760-44-7
  • Molecular Weight : 133.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1638760-44-7.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1638760-44-7.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart