AMBH303C56A9 Ambeed, Inc.

BRL 52537 hydrochloride

  • MDL number MFCD00672679
  • Cas Number : 112282-24-3
  • Molecular Weight : 391.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(3,4-Dichlorophenyl)-1-(2-(pyrrolidin-1-ylmethyl)piperidin-1-yl)ethanone hydrochloride
Purity 95
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/112282-24-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/112282-24-3.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart