AMBH303C56BE Ambeed, Inc.

Alvelestat

  • MDL number MFCD26960961
  • Cas Number : 848141-11-7
  • Molecular Weight : 545.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Methyl-5-(1-methyl-1H-pyrazol-5-yl)-N-((5-(methylsulfonyl)pyridin-2-yl)methyl)-2-oxo-1-(3-(trifluoromethyl)phenyl)-1,2-dihydropyridine-3-carboxamide
Purity 98
Storage temp. Sealed in dry,Store in freezer, under -20
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/848141-11-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/848141-11-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart