AMBH303C56D7 Ambeed, Inc.

(R)-tert-Butyl (1,4-dihydroxybutan-2-yl)carbamate

  • MDL number MFCD08703679
  • Cas Number : 397246-14-9
  • Molecular Weight : 205.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64-68°C
Purity 97
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 365°C at 760 mmHg
Product Link https://www.ambeed.com/products/397246-14-9.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/397246-14-9.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart