AMBH303C56D8 Ambeed, Inc.

(R)-4-tert-Butyl 3-methyl morpholine-3,4-dicarboxylate

  • MDL number MFCD09953091
  • Cas Number : 885321-46-0
  • Molecular Weight : 245.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/885321-46-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/885321-46-0.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart