AMBH303C56DC Ambeed, Inc.

5-(4-Chlorophenyl)-1-methyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD10006412
  • Cas Number : 1017414-83-3
  • Molecular Weight : 235.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1017414-83-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1017414-83-3.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart