AMBH303C56EA Ambeed, Inc.

2,4-Difluoro-6-methylpyridine

  • MDL number MFCD16610831
  • Cas Number : 83261-73-8
  • Molecular Weight : 129.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 139.0�0�t 760 mmHg
Product Link https://www.ambeed.com/products/83261-73-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/83261-73-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart