AMBH303C56EF Ambeed, Inc.

2-((4-Chlorophenyl)(piperidin-4-yloxy)methyl)pyridine 4-nitrobenzoate

  • MDL number MFCD19105231
  • Cas Number : 161558-45-8
  • Molecular Weight : 469.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/161558-45-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/161558-45-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart