AMBH303C56F4 Ambeed, Inc.

5-Chloro-6-methyluracil

  • MDL number MFCD00466419
  • Cas Number : 16018-87-4
  • Molecular Weight : 160.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 407.1°C at 760 mmHg
Product Link https://www.ambeed.com/products/16018-87-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/16018-87-4.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart