AMBH303C56F9 Ambeed, Inc.

Bicyclo[3.2.0]hept-2-en-6-one

  • MDL number MFCD00066201
  • Cas Number : 13173-09-6
  • Molecular Weight : 108.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Flash Point 66.3
Storage temp. Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 186.6�t 760 mmHg
Product Link https://www.ambeed.com/products/13173-09-6.html

Safety Information

signalword Danger
pcodes P261-P305+P351+P338
hcodes 3
MSDS https://www.ambeed.com/products/13173-09-6.html
pictograms GHS02,GHS07
hstatements H226-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart