AMBH303C571B Ambeed, Inc.

(Z)-Methyl 2-(hydroxyimino)-2-(pyridin-2-yl)acetate

  • MDL number MFCD27966261
  • Cas Number : 154410-82-9
  • Molecular Weight : 180.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/154410-82-9.html

Safety Information

signalword Warning
pcodes P264-P270-P301+P312-P330
MSDS https://www.ambeed.com/products/154410-82-9.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart