AMBH303C571C Ambeed, Inc.

Mephenesin

  • MDL number MFCD00004718
  • Cas Number : 59-47-2
  • Molecular Weight : 182.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/59-47-2.html

Safety Information

signalword Warning
pcodes P264-P270-P301+P312+P330-P501
MSDS https://www.ambeed.com/products/59-47-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart