AMBH303C5760 Ambeed, Inc.

(S)-3-(1-Aminoethyl)aniline

  • MDL number MFCD06761850
  • Cas Number : 317830-29-8
  • Molecular Weight : 136.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 135.2
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 266�t 760 mmHg
Product Link https://www.ambeed.com/products/317830-29-8.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/317830-29-8.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart