AMBH303C57A9 Ambeed, Inc.

Acalisib

  • MDL number N/A
  • Cas Number : 870281-34-8
  • Molecular Weight : 401.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (S)-2-(1-((9H-purin-6-yl)amino)ethyl)-6-fluoro-3-phenylquinazolin-4(3H)-one
Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/870281-34-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/870281-34-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart