AMBH303C57AD Ambeed, Inc.

Ethyl 4-chloroquinoline-6-carboxylate

  • MDL number MFCD16250588
  • Cas Number : 148018-34-2
  • Molecular Weight : 235.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/148018-34-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/148018-34-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart