AMBH303C57B0 Ambeed, Inc.

(6-Bromo-1-methyl-1,6-dihydropyridin-2-yl)methanamine

  • MDL number MFCD08690325
  • Cas Number : 675109-37-2
  • Molecular Weight : 203.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/675109-37-2.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/675109-37-2.html
pictograms GHS07
hstatements H302-H312-H332
Availability details for
Quantity
  • Availability
  • Add to Cart