AMBH303C57BC Ambeed, Inc.

2-(3-Bromophenoxy)-N,N-dimethylethanamine

  • MDL number MFCD03993547
  • Cas Number : 221915-84-0
  • Molecular Weight : 244.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Flash Point 123.1°C
Storage temp. Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Normal
Country of Origin China
Boiling point 280.0°20.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/221915-84-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/221915-84-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart