AMBH303C57BD Ambeed, Inc.

3-(2-Chloro-2-(4-chlorophenyl)vinyl)-2,2-dimethylcyclopropanecarboxylic acid

  • MDL number MFCD22421621
  • Cas Number : 88419-72-1
  • Molecular Weight : 285.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/88419-72-1.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/88419-72-1.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart