AMBH303C57E6 Ambeed, Inc.

2-Chloro-7-methyl-7H-purin-6-ol

  • MDL number MFCD00047225
  • Cas Number : 16017-76-8
  • Molecular Weight : 184.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-7-methyl-1H-purin-6(7H)-one
Purity 95
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 456.2°C at 760 mmHg
Product Link https://www.ambeed.com/products/16017-76-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/16017-76-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart